Chlorsulfuron
Chlorsulfuron
Katalog # EHERC11610000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2Cl)n1
- Inchi: InChI=1S/C12H12ClN5O4S/c1-7-14-10(17-12(15-7)22-2)16-11(19)18-23(20,21)9-6-4-3-5-8(9)13/h3-6H,1-2H3,(H2,14,15,16,17,18,19)
- IUPAC: 1-(2-chlorophenyl)sulfonyl-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:Chlorsulfuron
- UN No.:3077
- Cat. No.:EHERC11610000
- CAS-nr:64902-72-3
- Molekylvægt:357,77
- Fareklassebeskrivelse:GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):2-Chloro-N-[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]benzenesulfonamide;Chlorsulfuron;DPX 4189;DPX-W 4189;Glean;Glean 20DF;Glean 75;Glean 75DF;Kartes;Khardin;Kortes;Luhuanglong;N-(2-Chlorophenyl)sulfonyl-N'-(4-methyl-6-methoxy-2-triazinyl)urea;Tuligen;W 4189
- Shipping conditions:Room temperature
- Formel:C₁₂H₁₂ClN₅O₄S



