beta-Cyfluthrin
beta-Cyfluthrin
Katalog # EHERC11850200
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CC1(C)C(C=C(Cl)Cl)[C@@H]1C(=O)O[C@@H](C#N)c2ccc(F)c(Oc3ccccc3)c2
- Inchi: InChI=1S/C22H18Cl2FNO3/c1-22(2)15(11-19(23)24)20(22)21(27)29-18(12-26)13-8-9-16(25)17(10-13)28-14-6-4-3-5-7-14/h3-11,15,18,20H,1-2H3/t15?,18-,20+/m0/s1
- IUPAC: [(R)-cyano-(4-fluoro-3-phenoxyphenyl)methyl] (1S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
- Packing group: II
Specifikationer
Specifikationer
- Beskrivelse:beta-Cyfluthrin
- UN No.:3349
- Cat. No.:EHERC11850200
- CAS-nr:1820573-27-0
- Molekylvægt:434,29
- Fareklassebeskrivelse:GHS06, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):Baythroid XL;Beta-Baythroid;Beta-cyfluthrin;Bulldock;Bulldock 025EC;Bulldock 125SC;FCR 4545;Responsar;Tempo SC Ultra;Tempo Ultra;β-Cyfluthrin;(1S)-rel-3-(2,2-dichloroethenyl)-2,2-dimethyl-, (R)-cyano(4-fluoro-3-phenoxyphenyl)methyl Cyclopropanecarboxylic acid ester
- Shipping conditions:RT
- Formel:C₂₂H₁₈Cl₂FNO₃



