2-NP-AHD
2-NP-AHD
EHERC15654450
- SMILES: [O-][N+](=O)c1ccccc1\C=N\N2C(=O)CNC2=O
- Inchi: InChI=1S/C10H8N4O4/c15-9-6-11-10(16)13(9)12-5-7-3-1-2-4-8(7)14(17)18/h1-5H,6H2,(H,11,16)/b12-5+
- IUPAC: 3-[(E)-(2-nitrophenyl)methylideneamino]imidazolidine-2,4-dione
- Beskrivelse:2-NP-AHD
- Cat. No.:EHERC15654450
- CAS-nr:623145-57-3
- Molekylvægt:248,19
- Fareklassebeskrivelse:GHS07, GHS08
- Opbevaring betingelser:4 °C
- GHS signal:Farlig
- Synonym(s):1-[[(2-Nitrophenyl)methylene]amino]-2,4-imidazolidinedione
- Shipping conditions:RT
- Formel:C₁₀H₈N₄O₄