Perylene D12
Perylene D12
Catalog # EHERC20915100
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: [2H]c1c([2H])c2c([2H])c([2H])c([2H])c3c4c([2H])c([2H])c([2H])c5c([2H])c([2H])c([2H])c(c(c1[2H])c23)c45 Inchi: InChI=1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D IUPAC: 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuterioperylene
- Packing group: III
Specifications
Specifications
- Beskrivelse:Perylene D12
- UN No.:3077
- Cat. No.:EHERC20915100
- CAS-nr:1520-96-3
- Molekylvægt:264,38
- Fareklassebeskrivelse:GHS09
- Opbevaring betingelser:20 ℃
- GHS signal:Advarsel
- Synonym(s):Perylene-1,2,3,4,5,6,7,8,9,10,11,12-d12; Perylene-d12 (7CI,8CI,9CI); Perdeuterioperylene; [2H12]Perylene
- Shipping conditions:RT
- Formel:C₂₀₂H₁₂



