Coumatetralyl
Coumatetralyl
Katalog # EHERC11740000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: OC1=C(C2CCCc3ccccc23)C(=O)Oc4ccccc14 Inchi: InChI=1S/C19H16O3/c20-18-15-9-3-4-11-16(15)22-19(21)17(18)14-10-5-7-12-6-1-2-8-13(12)14/h1-4,6,8-9,11,14,20H,5,7,10H2 IUPAC: 4-hydroxy-3-(1,2,3,4-tetrahydronaphthalen-1-yl)chromen-2-one
- Packing group: I
Specifikationer
Specifikationer
- Beskrivelse:Coumatetralyl
- UN No.:2811
- Cat. No.:EHERC11740000
- CAS-nr:5836-29-3
- Molekylvægt:292,33
- Fareklassebeskrivelse:GHS06, GHS08, GHS09
- Opbevaring betingelser:20 ℃
- GHS signal:Farlig
- Synonym(s):2H-1-Benzopyran-2-one, 4-hydroxy-3-(1,2,3,4-tetrahydro-1-naphthalenyl)-; Coumarin, 4-hydroxy-3-(1,2,3,4-tetrahydro-1-naphthyl)- (6CI,7CI,8CI); 3-(1,2,3,4-Tetrahydro-1-naphthyl)-4-hydroxycoumarin; 4-Hydroxy-3-(1,2,3,4-tetrahydro-1-naphthyl)coumarin; Coumatetralyl; Endrocide; Murisan A; Racumin; Racumin 57; Rodentin; 3-(alpha-Tetralinyl)-4-hydroxycoumarin; 3-(alpha-Tetralyl)-4-Hydroxycoumarin
- Shipping conditions:RT
- Formel:C₁₉H₁₆O₃
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



