4-Chlorophenol
4-Chlorophenol
Katalog # EHERC11472000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Oc1ccc(Cl)cc1 Inchi: InChI=1S/C6H5ClO/c7-5-1-3-6(8)4-2-5/h1-4,8H IUPAC: 4-chlorophenol
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:4-Chlorophenol
- UN No.:2020
- Cat. No.:EHERC11472000
- CAS-nr:106-48-9
- Molekylvægt:128,56
- Fareklassebeskrivelse:GHS05, GHS06, GHS07, GHS09
- Opbevaring betingelser:20 ℃
- GHS signal:Farlig
- Synonym(s):Phenol, 4-chloro-; Phenol, p-chloro- (8CI); 4-Chlorophenol; 4-CP; 4-Hydroxychlorobenzene; Applied 3-78; NSC 2877; p-Chlorophenic acid; p-Chlorophenol
- Shipping conditions:RT
- Formel:C₆H₅ClO
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



