2,5-Dichlorophenol
2,5-Dichlorophenol
Katalog # EHERC12452000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Oc1cc(Cl)ccc1Cl Inchi: InChI=1S/C6H4Cl2O/c7-4-1-2-5(8)6(9)3-4/h1-3,9H IUPAC: 2,5-dichlorophenol
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:2,5-Dichlorophenol
- UN No.:3077
- Cat. No.:EHERC12452000
- CAS-nr:583-78-8
- Molekylvægt:163
- Fareklassebeskrivelse:GHS07, GHS08, GHS09
- Opbevaring betingelser:20 ℃
- GHS signal:Farlig
- Synonym(s):Phenol, 2,5-dichloro-; 2,5-Dichlorophenol; NSC 6296
- Shipping conditions:RT
- Formel:C₆H₄Cl₂O



