2,4-D-methyl ester
2,4-D-methyl ester
Katalog # EHERC11945000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: COC(=O)COc1ccc(Cl)cc1Cl Inchi: InChI=1S/C9H8Cl2O3/c1-13-9(12)5-14-8-3-2-6(10)4-7(8)11/h2-4H,5H2,1H3 IUPAC: methyl 2-(2,4-dichlorophenoxy)acetate
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:2,4-D-methyl ester
- UN No.:3077
- Cat. No.:EHERC11945000
- CAS-nr:1928-38-7
- Molekylvægt:235,06
- Fareklassebeskrivelse:GHS05, GHS07, GHS09
- Opbevaring betingelser:20 ℃
- GHS signal:Advarsel
- Shipping conditions:RT
- Formel:C₉H₈Cl₂O₃



