2,4'-DDD
2,4'-DDD
EHERC12030000
- SMILES: ClC1=CC=C(C=C1)C(C=2C=CC=CC2Cl)C(Cl)Cl Inchi: InChI=1S/C14H10Cl4/c15-10-7-5-9(6-8-10)13(14(17)18)11-3-1-2-4-12(11)16/h1-8,13-14H IUPAC: 1-chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene
- Beskrivelse:2,4'-DDD
- Cat. No.:EHERC12030000
- CAS-nr:53-19-0
- Molekylvægt:320,04
- Fareklassebeskrivelse:GHS07, GHS08
- Opbevaring betingelser:20 ℃
- GHS signal:Farlig
- Synonym(s):2,4'-DDD ;o,p'-DDD; CB-313; Lysodren; 1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane
- Shipping conditions:RT
- Formel:C₁₄H₁₀Cl₄



