2,3-Dichlorophenol
2,3-Dichlorophenol
Katalog # EHERC12450000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Oc1cccc(Cl)c1Cl Inchi: InChI=1S/C6H4Cl2O/c7-4-2-1-3-5(9)6(4)8/h1-3,9H IUPAC: 2,3-dichlorophenol
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:2,3-Dichlorophenol
- UN No.:2020
- Cat. No.:EHERC12450000
- CAS-nr:576-24-9
- Molekylvægt:163
- Fareklassebeskrivelse:GHS07, GHS08, GHS09
- Opbevaring betingelser:20 ℃
- GHS signal:Farlig
- Synonym(s):Phenol, 2,3-dichloro-; 2,3-Dichlorophenol; NSC 60646
- Shipping conditions:Room temperature
- Formel:C₆H₄Cl₂O
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



