Triphenylene
Triphenylene
Catalog # EHERC20945000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: c1ccc2c3ccccc3c4ccccc4c2c1
- Inchi: InChI=1S/C18H12/c1-2-8-14-13(7-1)15-9-3-4-11-17(15)18-12-6-5-10-16(14)18/h1-12H
- IUPAC: triphenylene
- Packing group: III
Specifications
Specifications
- Beskrivelse:Triphenylene
- UN No.:3077
- Cat. No.:EHERC20945000
- CAS-nr:217-59-4
- Molekylvægt:228,29
- Fareklassebeskrivelse:GHS05, GHS07, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):Triphenylene;1,2,3,4-Dibenznaphthalene;9,10-Benzophenanthrene;9,10-Benzphenanthrene;Benzo[l]phenanthrene;Isochrysene;NSC 57455
- Shipping conditions:RT
- Formel:C₁₈H₁₂
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



