Quinocetone
Quinocetone
Katalog # EHERC16709000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Cc1c(C(=O)\C=C\c2ccccc2)[n+]([O-])c3ccccc3[n+]1[O-]
- Inchi: InChI=1S/C18H14N2O3/c1-13-18(17(21)12-11-14-7-3-2-4-8-14)20(23)16-10-6-5-9-15(16)19(13)22/h2-12H,1H3/b12-11+
- IUPAC: (E)-1-(3-methyl-1,4-dioxidoquinoxaline-1,4-diium-2-yl)-3-phenylprop-2-en-1-one
Specifikationer
Specifikationer
- Beskrivelse:Quinocetone
- Cat. No.:EHERC16709000
- CAS-nr:81810-66-4
- Molekylvægt:306,32
- Fareklassebeskrivelse:GHS07, GHS08
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):2-Propen-1-one;1-(3-methyl-2-quinoxalinyl)-3-phenyl-;N,N'-dioxide;1-(3-Methyl-1,4-dioxido-2-quinoxalinyl)-3-phenyl-2-propen-1-one;NSC 621477;Quinocetone
- Shipping conditions:RT
- Formel:C₁₈H₁₄N₂O₃
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



