Oxamyl
Oxamyl
EHERC15780000
- SMILES: CNC(=O)O\N=C(/SC)\C(=O)N(C)C
- Inchi: InChI=1S/C7H13N3O3S/c1-8-7(12)13-9-5(14-4)6(11)10(2)3/h1-4H3,(H,8,12)/b9-5-
- IUPAC: methyl (1Z)-2-(dimethylamino)-N-(methylcarbamoyloxy)-2-oxoethanimidothioate
- Packing group: I
- Beskrivelse:Oxamyl
- UN No.:2757
- Cat. No.:EHERC15780000
- CAS-nr:23135-22-0
- Molekylvægt:219,26
- Fareklassebeskrivelse:GHS06, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):Oxamyl;2-(Dimethylamino)-N-[[(methylamino)carbonyl]oxy]-2-oxo-ethanimidothioic Acid Methyl Ester;N',N'-Dimethyl-N-[(methylcarbamoyl)oxy]-1-thio-Oxamimidic Acid Methyl Ester;Methylcarbamic acid O-[[[(dimethylcarbamoyl)methylthio]methylene]amino] deriv.;DPX 1410;DPX 1410L;Du Pont 1410;NSC 379588;Oxamyl (pesticide);Thioxamyl;Vydate;Vydate CLV;Vydate G;Vydate L
- Shipping conditions:RT
- Formel:C₇H₁₃N₃O₃S



