Hexaconazole
Hexaconazole
EHERC14190000
- SMILES: CCCCC(O)(Cn1cncn1)c2ccc(Cl)cc2Cl
- Inchi: InChI=1S/C14H17Cl2N3O/c1-2-3-6-14(20,8-19-10-17-9-18-19)12-5-4-11(15)7-13(12)16/h4-5,7,9-10,20H,2-3,6,8H2,1H3
- IUPAC: 2-(2,4-dichlorophenyl)-1-(1,2,4-triazol-1-yl)hexan-2-ol
- Packing group: III
- Beskrivelse:Hexaconazole
- UN No.:3077
- Cat. No.:EHERC14190000
- CAS-nr:79983-71-4
- Molekylvægt:314,21
- Fareklassebeskrivelse:GHS07, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):1H-1,2,4-Triazole-1-ethanol;α-butyl-α-(2,4-dichlorophenyl)-;(±)-;α-Butyl-α-(2,4-dichlorophenyl)-1H-1,2,4-triazole-1-ethanol;(±)-Hexaconazole;2-(2,4-Dichlorophenyl)-1-(1H-1,2,4-triazol-1-yl)hexan-2-ol;Anvil;Anvil L;Anvil Liquid;Blin Exa;Canvil;Clortriafol;Contaf;Contaf 5EC;Contaf Plus;Controll;FD 4053;Flowmax 5SC;Hexaconazole;Hexaconazole Technical;Hexadhan;PC 1002;PP 523;R 154523;RS-2-(2,4-Dichlorophenyl)-1-(1H-1,2,4-triazol-1-yl)hexan-2-ol;Ranvil;Samarth;Sitara;TDP;1H-1,2,4-Triazole-1-ethanol;α-butyl-α-(2,4-dichlorophenyl)-
- Shipping conditions:RT
- Formel:C₁₄H₁₇Cl₂N₃O



