Dichlofluanid
Dichlofluanid
Katalog # EHERC12300000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CN(C)S(=O)(=O)N(SC(F)(Cl)Cl)c1ccccc1
- Inchi: InChI=1S/C9H11Cl2FN2O2S2/c1-13(2)18(15,16)14(17-9(10,11)12)8-6-4-3-5-7-8/h3-7H,1-2H3
- IUPAC: N-[dichloro(fluoro)methyl]sulfanyl-N-(dimethylsulfamoyl)aniline
- Packing group: II
Specifikationer
Specifikationer
- Beskrivelse:Dichlofluanid
- UN No.:2761
- Cat. No.:EHERC12300000
- CAS-nr:1085-98-9
- Molekylvægt:333,23
- Fareklassebeskrivelse:GHS06, GHS07, GHS09
- Opbevaring betingelser:4 °C
- GHS signal:Farlig
- Synonym(s):Sulfamide;N-[(dichlorofluoromethyl)thio]-N',N'-dimethyl-N-phenyl- (7CI,8CI);1,1-Dichloro-N-[(dimethylamino)sulfonyl]-1-fluoro-N-phenylmethanesulfenamide;BAY 47531;Bayer 47531;Chlormethionate;Coatcide TW;Dichlofluanid;Dichlofluanide;Diclofluanide;Elvaron;Eparen;Euparen;Euparen WG;Lichenicide 246;N,N-Dimethyl-N'-(dichlorofluoromethylthio)-N'-phenylsulfamide;N,N-Dimethyl-N'-phenyl-N'-(dichlorofluoromethylthio)sulfamide;N,N-Dimethyl-N'-phenyl-N'-(fluorodichloromethylthio)sulfamide;N-Phenyl-N-(fluorodichloromethylsulfenyl)-N',N'-dimethylsulfamide;N-[(Dichlorofluoromethyl)thio]-N',N'-dimethyl-N-phenylsulfamide;NSC 218451;N'-(Dichlorofluoromethylthio)-N,N-dimethyl-N'-phenylsulfamide;Oiparen;Pecudin;Preventol A 4;Preventol A 4S
- Shipping conditions:RT
- Formel:C₉H₁₁Cl₂FN₂O₂S₂
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



