Desmedipham
Desmedipham
Katalog # EHERC12160000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CCOC(=O)Nc1cccc(OC(=O)Nc2ccccc2)c1
- Inchi: InChI=1S/C16H16N2O4/c1-2-21-15(19)18-13-9-6-10-14(11-13)22-16(20)17-12-7-4-3-5-8-12/h3-11H,2H2,1H3,(H,17,20)(H,18,19)
- IUPAC: [3-(ethoxycarbonylamino)phenyl] N-phenylcarbamate
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:Desmedipham
- UN No.:3077
- Cat. No.:EHERC12160000
- CAS-nr:13684-56-5
- Molekylvægt:300,31
- Fareklassebeskrivelse:GHS08, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):Carbamic acid;[3-[[(phenylamino)carbonyl]oxy]phenyl]-;ethyl ester (9CI);Carbanilic acid;m-hydroxy-;ethyl ester;carbanilate (ester) (8CI);3-Ethoxycarbonylaminophenyl N-phenylcarbamate;Betanex;DMP Stefes;Desmedipham;Destor;EP 475;EP 475 (pesticide);Ethyl N-[3-(N-phenylcarbamoyloxy)phenyl]carbamate;Ethyl m-hydroxycarbanilate carbanilate;SN 38107;Synbetan D
- Shipping conditions:RT
- Formel:C₁₆H₁₆N₂O₄
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



