DDT (technical)
DDT (technical)
Catalog # EHERC12080000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: ClC1=CC=C(C(C2=CC=CC=C2Cl)C(Cl)(Cl)Cl)C=C1.ClC3=CC=C(C(C4=CC=C(Cl)C=C4)C(Cl)(Cl)Cl)C=C3
- Inchi: InChI=1S/2C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10;15-10-7-5-9(6-8-10)13(14(17,18)19)11-3-1-2-4-12(11)16/h2*1-8,13H
- IUPAC: 1-chloro-2-(2,2,2-trichloro-1-(4-chlorophenyl)ethyl)benzene--4,4'-(2,2,2-trichloroethane-1,1-diyl)bis(chlorobenzene) (1/1)
- Packing group: III
Specifications
Specifications
- Beskrivelse:DDT (technical)
- UN No.:2761
- Cat. No.:EHERC12080000
- CAS-nr:8017-34-3
- Molekylvægt:708,98
- Fareklassebeskrivelse:GHS06, GHS08, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):1,1'-(2,2,2-Trichloroethylidene)bis[4-chloro-benzene;1-Chloro-2-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]-benzene Mixture With 1,1'-(2,2,2-Trichloroethylidene)bis[4-chlorobenzene];
- Shipping conditions:RT
- Formel:C₁₄H₉Cl₅•(C₁₄H₉Cl₅)



