D-alpha-Tocopherol
D-alpha-Tocopherol
Katalog # EHERCA17924200
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(O)c(C)c(C)c2O1
- Inchi: InChI=1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21-,22-,29-/m1/s1
- IUPAC: (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
Specifikationer
Specifikationer
- Beskrivelse:D-alpha-Tocopherol
- Cat. No.:EHERCA17924200
- CAS-nr:59-02-9
- Molekylvægt:430,71
- Opbevaring betingelser:–18 °C
- Synonym(s):RRR-alpha-Tocopherol;(2R)-2,5,7,8-Tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol;(2R)-3,4-dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-Benzopyran-6-ol;2H-1-Benzopyran-6-ol;3,4-dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-;[2R-[2R*(4R*,8R*)]]-;(+)-2R,4'R,8'R-α-Tocopherol;(+)-α-Tocopherol;(2R)-3,4-Dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-ol;(2R,4'R,8'R)-α-Tocopherol;(R,R,R)-α-Tocopherol;(all-R)-α-Tocopherol;5,7,8-Trimethyltocol;Almefrol;Biopass E 20;Cadvion;Cardi-E;Copherol F 1300C;Covitol F 1000;Covitol F 1000-2;Covitol F 1300;Covitol F 1370;Covitol IK 00069;D-α-Tocopherol;Denamone;E 307;E 307 (tocopherol);E 5-87;E-Oil 1000;EMF 1490;Emipherol;Endo E;Eprolin;Eprolin S;Epsilan;Esorb;Etamican;Etavit;Evitaminum;Ilitia;Irganox E 201;NSC 20812;NatAlc;Natur-E micelle;Phytogermin;Profecundin;Rhenogran Ronotec 50;Ronotec 2001;Spavit E;Syntopherol;Tenox GT 1;Tokopharm;Vascuals;Verrol;Vitamin Eα;Vitaplex E;Vitayonon;Viteolin;d-α-Tocopherol;α-D-Tocopherol;α-Tocopherol;α-Vitamin E
- Shipping conditions:20 °C
- Formel:C₂₉H₅₀O₂
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



