Cyclohexanone
Cyclohexanone
Katalog # EHERC11825500
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: O=C1CCCCC1
- Inchi: InChI=1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2
- IUPAC: cyclohexanone
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:Cyclohexanone
- UN No.:1915
- Cat. No.:EHERC11825500
- CAS-nr:108-94-1
- Molekylvægt:98,14
- Fareklassebeskrivelse:GHS02, GHS05, GHS06, GHS07
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):Cyclohexanone;Anon;Anone;C 0489;Hexanon;Hytrol O;NSC 5711;Nadone;Pimelic ketone;Pimelin ketone;Sextone
- Shipping conditions:RT
- Formel:C₆H₁₀O
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



