Creatine
Creatine
Katalog # EHERC11748500
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CN(CC(=O)O)C(=N)N
- Inchi: InChI=1S/C4H9N3O2/c1-7(4(5)6)2-3(8)9/h2H2,1H3,(H3,5,6)(H,8,9)
- IUPAC: 2-[carbamimidoyl(methyl)amino]acetic acid
Specifikationer
Specifikationer
- Beskrivelse:Creatine
- Cat. No.:EHERC11748500
- CAS-nr:57-00-1
- Molekylvægt:131,13
- Opbevaring betingelser:20 °C
- Synonym(s):Creatine;N-(Aminoiminomethyl)-N-methylglycine;2-(1-Methylguanidino)acetic Acid;Cosmocair C 100;Kre-Alkalyn;Methylguanidoacetic Acid;N-Methyl-N-guanylglycine;NSC 8752;Neotine;Phosphagen;Tego Cosmo C 100;[N-(Amidino)-N-methylamino]acetic Acid;
- Shipping conditions:RT
- Formel:C₄H₉N₃O₂
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



