Chlorpropham
Chlorpropham
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CC(C)OC(=O)Nc1cccc(Cl)c1
- Inchi: InChI=1S/C10H12ClNO2/c1-7(2)14-10(13)12-9-5-3-4-8(11)6-9/h3-7H,1-2H3,(H,12,13)
- IUPAC: propan-2-yl N-(3-chlorophenyl)carbamate
- Packing group: III
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Beskrivelse:Chlorpropham
- UN No.:3077
- Cat. No.:EHERC11580000
- CAS-nr:101-21-3
- Molekylvægt:213,66
- Fareklassebeskrivelse:GHS07, GHS08, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):Carbamic acid;(3-chlorophenyl)-;1-methylethyl ester (9CI);Carbanilic acid;m-chloro-;isopropyl ester (6CI,7CI,8CI);Bud Nip;Bygran;CI-IPC;CIPC;Chlor IFC;Chlor IFK;Chlor IPC;Chloro-IPC;Chloropropham;Chlorpropham;Cl-IFK;Elbanil;Furloe;Furloe 3EC;IPCPC;Isopropyl 3-chlorocarbanilate;Isopropyl N-(3-chlorophenyl)carbamate;Isopropyl N-(m-chlorophenyl)carbamate;Isopropyl m-chlorocarbanilate;Keim-Stop;Liro CIPC;Metoxon;Mirvale;NSC 29466;Neo Stop 5;Nexoval;Preventol;Preventol 56;Shield Potato Sprout Inhibitor;Triherbicide CIPC;Y 3;m-Chlorocarbanilic acid isopropyl ester
- Shipping conditions:RT
- Formel:C₁₀H₁₂ClNO₂
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



