4,4'-DDMU
4,4'-DDMU
Katalog # EHERC12061000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: ClC=C(c1ccc(Cl)cc1)c2ccc(Cl)cc2
- Inchi: InChI=1S/C14H9Cl3/c15-9-14(10-1-5-12(16)6-2-10)11-3-7-13(17)8-4-11/h1-9H
- IUPAC: 1-chloro-4-[2-chloro-1-(4-chlorophenyl)ethenyl]benzene
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:4,4'-DDMU
- UN No.:3077
- Cat. No.:EHERC12061000
- CAS-nr:1022-22-6
- Molekylvægt:283,58
- Fareklassebeskrivelse:GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):Benzene;1,1'-(chloroethenylidene)bis[4-chloro- (9CI);Ethylene;2-chloro-1,1-bis(p-chlorophenyl)- (6CI,7CI,8CI);1,1'-(2-Chloroethenylidene)bis[4-chlorobenzene];1,1-Bis(4-chlorophenyl)-2-chloroethene;1,1-Bis(4-chlorophenyl)-2-chloroethylene;1,1-Bis(p-chlorophenyl)-2-chloroethene;1,1-Bis(p-chlorophenyl)-2-chloroethylene;1,1'-(Chlorovinylidene)bis[4-chlorobenzene];1-Chloro-2,2-bis(4-chlorophenyl)ethylene;1-Chloro-2,2-bis(p-chlorophenyl)ethylene;2,2-Bis(4-chlorophenyl)-1-chloroethylene;2,2-Bis(p-chlorophenyl)-1-chloroethylene;2-Chloro-1,1-bis(4-chlorophenyl)ethylene;DDMU;NSC 46465;TDEE;p,p'-1-Chloro-2,2'-bis(4-chlorophenyl)ethene;p,p'-DDD olefin;p,p'-DDM;p,p'-DDMU;p,p'-DDMU;p,p'-DME;p,p'-TDE olefin;p,p'-TDEE
- Shipping conditions:RT
- Formel:C₁₄H₉Cl₃
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



