3-Chlorophenol
3-Chlorophenol
Katalog # EHERC11471000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Oc1cccc(Cl)c1
- Inchi: InChI=1S/C6H5ClO/c7-5-2-1-3-6(8)4-5/h1-4,8H
- IUPAC: 3-chlorophenol
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:3-Chlorophenol
- UN No.:2020
- Cat. No.:EHERC11471000
- CAS-nr:108-43-0
- Molekylvægt:128,56
- Fareklassebeskrivelse:GHS05, GHS07, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):Phenol;3-chloro-;Phenol;m-chloro- (7CI,8CI);3-Chlorophenol;3-Hydroxychlorobenzene;NSC 59700;m-Chlorophenic acid;m-Chlorophenol
- Shipping conditions:RT
- Formel:C₆H₅ClO



