2-Nitrophenol
2-Nitrophenol
Katalog # EHERC15590200
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Oc1ccccc1[N+](=O)[O-]
- Inchi: InChI=1S/C6H5NO3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H
- IUPAC: 2-nitrophenol
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:2-Nitrophenol
- UN No.:1663
- Cat. No.:EHERC15590200
- CAS-nr:88-75-5
- Molekylvægt:139,11
- Fareklassebeskrivelse:GHS07, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):Phenol;2-nitro-;Phenol;o-nitro- (8CI);2-Nitrophenol;2-Hydroxynitrobenzene;NSC 1552;o-Hydroxynitrobenzene;o-Nitrophenol
- Shipping conditions:RT
- Formel:C₆H₅NO₃
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



