2,3-Dinitrotoluene
2,3-Dinitrotoluene
EHERC12786100
- SMILES: Cc1cccc(c1[N+](=O)[O-])[N+](=O)[O-]
- Inchi: InChI=1S/C7H6N2O4/c1-5-3-2-4-6(8(10)11)7(5)9(12)13/h2-4H,1H3
- IUPAC: 1-methyl-2,3-dinitrobenzene
- Packing group: II
- Beskrivelse:2,3-Dinitrotoluene
- UN No.:3454
- Cat. No.:EHERC12786100
- CAS-nr:602-01-7
- Molekylvægt:182,13
- Fareklassebeskrivelse:GHS06, GHS08, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):1-Methyl-2,3-dinitrobenzene;Toluene;2,3-dinitro- (8CI);1-Methyl-2,3-dinitrobenzene;2,3-DNT;2,3-Dinitrotoluene;2,3-Dinitrotoluol
- Shipping conditions:RT
- Formel:C₇H₆N₂O₄
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



