Valeric acid-methyl ester
Valeric acid-methyl ester
EHERC17899750
- SMILES: CCCCC(=O)OC
- Inchi: InChI=1S/C6H12O2/c1-3-4-5-6(7)8-2/h3-5H2,1-2H3
- IUPAC: methyl pentanoate
- Packing group: II
- Beskrivelse:Valeric acid-methyl ester
- UN No.:3272
- Cat. No.:EHERC17899750
- CAS-nr:624-24-8
- Molekylvægt:116,16
- Fareklassebeskrivelse:GHS02, GHS07
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Shipping conditions:RT
- Formel:C₆H₁₂O₂



