Sulfamoxol
Sulfamoxol
Katalog # EHERC16998200
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Cc1oc(NS(=O)(=O)c2ccc(N)cc2)nc1C
- Inchi: InChI=1S/C11H13N3O3S/c1-7-8(2)17-11(13-7)14-18(15,16)10-5-3-9(12)4-6-10/h3-6H,12H2,1-2H3,(H,13,14)
- IUPAC: 4-amino-N-(4,5-dimethyl-1,3-oxazol-2-yl)benzenesulfonamide
Specifikationer
Specifikationer
- Beskrivelse:Sulfamoxol
- Cat. No.:EHERC16998200
- CAS-nr:729-99-7
- Molekylvægt:267,3
- Fareklassebeskrivelse:GHS07
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):Benzenesulfonamide, 4-amino-N-(4,5-dimethyl-2-oxazolyl)-; Sulfanilamide, N1-(4,5-dimethyl-2-oxazolyl)- (6CI,8CI); 2-(p-Aminobenzolsulfonamido)-4,5-dimethyloxazole; 2-Sulfanilamido-4,5-dimethyloxazole; Justamil; N1-(4,5-Dimethyl-2-oxazolyl)sulfanilamide; NSC 683535; N'-(4,5-Dimethyl-2-oxazolyl)sulfanilamide; Oxasulfa; Sulfabutin; Sulfadimethyloxazole; Sulfamoxole; Sulfamoxolum; Sulfano; Sulfavigor; Sulfmidil; Sulfono; Sulfune; Sulfuno; Tardamid; Tardamide
- Shipping conditions:RT
- Formel:C₁₁H₁₃N₃O₃S
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



