PCB No. 170
PCB No. 170
Katalog # EHERC20017000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Clc1ccc(c(Cl)c1Cl)c2cc(Cl)c(Cl)c(Cl)c2Cl
- Inchi: InChI=1S/C12H3Cl7/c13-6-2-1-4(8(15)10(6)17)5-3-7(14)11(18)12(19)9(5)16/h1-3H
- IUPAC: 1,2,3,4-tetrachloro-5-(2,3,4-trichlorophenyl)benzene
- Packing group: II
Specifikationer
Specifikationer
- Beskrivelse:PCB No. 170
- UN No.:3432
- Cat. No.:EHERC20017000
- CAS-nr:35065-30-6
- Molekylvægt:395,32
- Fareklassebeskrivelse:GHS08, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):1,1'-Biphenyl, 2,2',3,3',4,4',5-heptachloro-; 2,2',3,3',4,4',5-Heptachloro-1,1'-biphenyl; 2,2',3,3',4,4',5-Heptachlorobiphenyl; 2,3,4,2',3',4',5'-Heptachlorobiphenyl; 2,3,4,5,2',3',4'-Heptachlorobiphenyl; CB 170; PCB 170; 2,2',3,3',4,4',5-HeptaCB
- Shipping conditions:RT
- Formel:C₁₂H₃Cl₇
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



