N-(2-Hydroxyethyl)-N-methylaniline
N-(2-Hydroxyethyl)-N-methylaniline
EHERC14231560
- SMILES: CN(CCO)c1ccccc1
- Inchi: InChI=1S/C9H13NO/c1-10(7-8-11)9-5-3-2-4-6-9/h2-6,11H,7-8H2,1H3
- IUPAC: 2-(N-methylanilino)ethanol
- Beskrivelse:N-(2-Hydroxyethyl)-N-methylaniline
- Cat. No.:EHERC14231560
- CAS-nr:93-90-3
- Molekylvægt:151,21
- Fareklassebeskrivelse:GHS07
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):Ethanol, 2-(N-methylanilino)- (6CI,7CI,8CI); 2-(Methylphenylamino)ethanol; 2-(N-Methyl-N-phenylamino)ethanol; 2-(N-Methylanilino)ethanol; N-(2-Hydroxyethyl)-N-methylaniline; N-Hydroxyethyl-N-methylaniline; N-Methyl-N-(2-hydroxyethyl)aniline; N-Methyl-N-hydroethyl aniline; N-Methyl-N-phenyl-2-aminoethanol; N-Methyl-N-phenyl-2-hydroxyethylamine; N-Methyl-N-phenylethanolamine; N-Phenyl-N-methyl-ethanolamine; NSC 9274
- Shipping conditions:RT
- Formel:C₉H₁₃NO



