Mesterolone
Mesterolone
EHERC14914900
- SMILES: C[C@H]1CC(=O)C[C@@H]2CC[C@H]3[C@@H]4CC[C@H](O)[C@@]4(C)CC[C@@H]3[C@@]12C
- Inchi: InChI=1S/C20H32O2/c1-12-10-14(21)11-13-4-5-15-16-6-7-18(22)19(16,2)9-8-17(15)20(12,13)3/h12-13,15-18,22H,4-11H2,1-3H3/t12-,13-,15-,16-,17-,18-,19-,20-/m0/s1
- IUPAC: (1S,5S,8R,9S,10S,13S,14S,17S)-17-hydroxy-1,10,13-trimethyl-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
- Packing group: III
- Beskrivelse:Mesterolone
- UN No.:3077
- Cat. No.:EHERC14914900
- CAS-nr:1424-00-6
- Molekylvægt:304,47
- Fareklassebeskrivelse:GHS08, GHS09
- Opbevaring betingelser:4 °C
- GHS signal:Farlig
- Synonym(s):Androviron; MEST; Mesteranum; Mesterolone; Mestoranum; NSC 75054; Proviron; Provirone 25; SH 723; Testiwop; Androstan-3-one, 17-hydroxy-1-methyl-, (1alpha,5alpha,17beta)-; 5alpha-Androstan-3-one, 17beta-hydroxy-1alpha-methyl- (6CI,7CI,8CI); (1alpha,5alpha,17beta)-17-Hydroxy-1-methylandrostan-3-one; 17beta-Hydroxy-1alpha-methyl-5alpha-androstan-3-one; 1alpha-Methyl-17beta-hydroxy-5alpha-androstan-3-one; 1alpha-Methyl-5alpha-androstan-17beta-ol-3-one; 1alpha-Methyl-5alpha-dihydrotestosterone
- Shipping conditions:RT
- Formel:C₂₀H₃₂O₂



