Fenpropimorph
Fenpropimorph
Katalog # EHERCA13540000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CC(CN1C[C@@H](C)O[C@@H](C)C1)Cc2ccc(cc2)C(C)(C)C
- Inchi: InChI=1S/C20H33NO/c1-15(12-21-13-16(2)22-17(3)14-21)11-18-7-9-19(10-8-18)20(4,5)6/h7-10,15-17H,11-14H2,1-6H3/t15?,16-,17+
- IUPAC: (2R,6S)-4-[3-(4-tert-butylphenyl)-2-methylpropyl]-2,6-dimethylmorpholine
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:Fenpropimorph
- UN No.:3082
- Cat. No.:EHERCA13540000
- CAS-nr:67564-91-4
- Molekylvægt:303,48
- Fareklassebeskrivelse:GHS07, GHS08, GHS09
- Opbevaring betingelser:–18 °C
- GHS signal:Farlig
- Synonym(s):Morpholine, 4-[3-[4-(1,1-dimethylethyl)phenyl]-2-methylpropyl]-2,6-dimethyl-, cis-; rel-(2R,6S)-4-[3-[4-(1,1-Dimethylethyl)phenyl]-2-methylpropyl]-2,6-dimethylmorpholine; BAS 42100F; Corbel; Fenpropimorph; Forbel 750; Mistral; Volley 88OL; cis-4-[3-(4-tert-Butylphenyl)-2-methylpropyl]-2,6-dimethylmorpholine
- Shipping conditions:RT
- Formel:C₂₀H₃₃NO



