Fenamiphos
Fenamiphos
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CCOP(=O)(NC(C)C)Oc1ccc(SC)c(C)c1
- Inchi: InChI=1S/C13H22NO3PS/c1-6-16-18(15,14-10(2)3)17-12-7-8-13(19-5)11(4)9-12/h7-10H,6H2,1-5H3,(H,14,15)
- IUPAC: N-[ethoxy-(3-methyl-4-methylsulfanylphenoxy)phosphoryl]propan-2-amine
- Packing group: II
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Beskrivelse:Fenamiphos
- UN No.:2783
- Cat. No.:EHERC13420000
- CAS-nr:22224-92-6
- Molekylvægt:303,36
- Fareklassebeskrivelse:GHS06, GHS07, GHS09
- Opbevaring betingelser:–18 °C
- GHS signal:Farlig
- Synonym(s):Phosphoramidic acid, (1-methylethyl)-, ethyl 3-methyl-4-(methylthio)phenyl ester (9CI); Phosphoramidic acid, isopropyl-, ethyl 4-(methylthio)-m-tolyl ester (8CI); B 68138; BAY 68138; Bayer 68138; Ethyl 3-methyl-4-(methylthio)phenyl (1-methylethyl)phosphoramidate; Ethyl 4-(methylthio)-m-tolyl isopropylphosphoramidate; Fenamifos; Fenamiphos; Fenamithion; Nemacur; Nemacur P; Phenamiphos; Phosphoramidic acid, N-(1-methylethyl)-, ethyl 3-methyl-4-(methylthio)phenyl ester
- Shipping conditions:RT
- Formel:C₁₃H₂₂NO₃PS



