Bioresmethrin
Bioresmethrin
EHERC10620000
- SMILES: CC(=C[C@@H]1[C@@H](C(=O)OCc2coc(Cc3ccccc3)c2)C1(C)C)C
- Inchi: InChI=1S/C22H26O3/c1-15(2)10-19-20(22(19,3)4)21(23)25-14-17-12-18(24-13-17)11-16-8-6-5-7-9-16/h5-10,12-13,19-20H,11,14H2,1-4H3/t19-,20+/m1/s1
- IUPAC: (5-benzylfuran-3-yl)methyl (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
- Packing group: III
- Beskrivelse:Bioresmethrin
- UN No.:3077
- Cat. No.:EHERC10620000
- CAS-nr:28434-01-7
- Molekylvægt:338,44
- Fareklassebeskrivelse:GHS07, GHS09
- Opbevaring betingelser:4 °C
- GHS signal:Advarsel
- Shipping conditions:RT
- Formel:C₂₂H₂₆O₃



