4,4'-DDT
4,4'-DDT
Katalog # EHERC12082000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Clc1ccc(cc1)C(c2ccc(Cl)cc2)C(Cl)(Cl)Cl
- Inchi: InChI=1S/C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10/h1-8,13H
- IUPAC: 1-chloro-4-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:4,4'-DDT
- UN No.:2761
- Cat. No.:EHERC12082000
- CAS-nr:50-29-3
- Molekylvægt:354,49
- Fareklassebeskrivelse:GHS06, GHS08, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):Benzene, 1,1'-(2,2,2-trichloroethylidene)bis[4-chloro-; Ethane, 1,1,1-trichloro-2,2-bis(p-chlorophenyl)- (7CI,8CI); 1,1'-(2,2,2-Trichloroethylidene)bis[4-chlorobenzene]; 1,1,1-Trichloro-2,2-bis(p-chlorophenyl)ethane; 1,1,1-Trichloro-2,2-di(p-chlorophenyl)ethane; 1,1,1-Trichloro-2-2,2-bis(p-chlorophenyl)ethane; 1,1-Bis(4-chlorophenyl)-2,2,2-trichloroethane; 1,1-Bis(p-chlorophenyl)-2,2,2-trichloroethane; 2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethane; 2,2-Bis(4-chlorophenyl)-1,1,1-trichloroethane; 2,2-Bis(p-chlorophenyl)-1,1,1-trichloroethane; 4,4'-DDT; 4,4'-Dichlorodiphenyltrichloroethane; Aavero-extra; Agritan; Arkotine; Azotox M 33; Benzochloryl; Bosan supra; Bovidermol; Chlorophenothane; Chlorphenothan; Chlorphenotoxum; Citox; Clofenotan; Clofenotane; DDT; Deoval; Detox; Detox (pesticide); Detoxan; Dibovin; Dichlorodiphenyltrichloroethane; Dicophane; Dicophaner; Dodat; Dykol; ENT-1506; Estonate; Ethane, 1,1,1-trichloro-2,2-bis(4-chlorophenyl)-; FWJ; Gesafid; Gesapon; Gesarex; Gesarol; Guesapon; Hildit; Ivoran; Mutoxan; NSC 8939; Neocid; Neocidol; Neocidol (solid); PEB1; Parachlorocidum; Pentachlorin; Penticidum; Tafidex; Trichlorobis(4'-chlorophenyl)ethane; Zerdane; o,p'-1,1,1-Trichloro-2-2,2-bis(p-chlorophenyl)ethane; p,p'-DDT; p,p'-Dichlorodiphenyltrichloroethane; p,p'-Dichlorodiphenyltrichloromethylmethane; α,α-Bis(p-chlorophenyl)-β,β,β-trichlorethane
- Shipping conditions:RT
- Formel:C₁₄H₉Cl₅



