4,4'-DDA
4,4'-DDA
Katalog # EHERC12020000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: OC(=O)C(c1ccc(Cl)cc1)c2ccc(Cl)cc2
- Inchi: InChI=1S/C14H10Cl2O2/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8,13H,(H,17,18)
- IUPAC: 2,2-bis(4-chlorophenyl)acetic acid
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:4,4'-DDA
- UN No.:3077
- Cat. No.:EHERC12020000
- CAS-nr:83-05-6
- Molekylvægt:281,13
- Fareklassebeskrivelse:GHS07, GHS08, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):Acetic acid, bis(p-chlorophenyl)- (6CI,7CI,8CI); 4-Chloro-α-(4-chlorophenyl)benzeneacetic acid; 2,2-Bis(4-chlorophenyl)acetic acid; Bis(4-chlorophenyl)acetic acid; Bis(p-chlorophenyl)acetic acid; DDA; DDA (degradation product); Di(p-chlorophenyl)acetic acid; Dichlorodiphenylacetic acid; NSC 4279; p,p'-DDA
- Shipping conditions:RT
- Formel:C₁₄H₁₀Cl₂O₂
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



