3,4-Dichlorophenol
3,4-Dichlorophenol
Katalog # EHERC12453400
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Oc1ccc(Cl)c(Cl)c1
- Inchi: InChI=1S/C6H4Cl2O/c7-5-2-1-4(9)3-6(5)8/h1-3,9H
- IUPAC: 3,4-dichlorophenol
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:3,4-Dichlorophenol
- UN No.:2020
- Cat. No.:EHERC12453400
- CAS-nr:95-77-2
- Molekylvægt:163
- Fareklassebeskrivelse:GHS07, GHS08, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):Phenol, 3,4-dichloro-; 3,4-Dichlorophenol; 4,5-Dichlorophenol; NSC 60648
- Shipping conditions:Room Temperature
- Formel:C₆H₄Cl₂O
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



