2,6-Dibromophenol
2,6-Dibromophenol
Katalog # EHERC12241200
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: Oc1c(Br)cccc1Br
- Inchi: InChI=1S/C6H4Br2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
- IUPAC: 2,6-dibromophenol
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:2,6-Dibromophenol
- UN No.:2811
- Cat. No.:EHERC12241200
- CAS-nr:608-33-3
- Molekylvægt:251,9
- Fareklassebeskrivelse:GHS06, GHS07
- Opbevaring betingelser:20 °C
- GHS signal:Farlig
- Synonym(s):Phenol, 2,6-dibromo-; 2,6-Dibromophenol; NSC 6214
- Shipping conditions:Room Temperature
- Formel:C₆H₄Br₂O



