Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Potassium acetate ≥98%

Potassium acetate ≥98%

Bottle
Katalog # APOSBUP19847-10G
New Product
Leverandør:   Apollo Scientific
Potassium acetate ≥98%
Katalog # APOSBUP19847-10G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  127-08-2
Potassium acetate ≥98%

Bottle

CAS nummer:  127-08-2
Price
659,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonymer:  Acetic acid potassium salt
  • CAS nummer:127-08-2
  • Formel:C₂H₃KO₂
  • Molecular Weight:98,14 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer