2-Bromo-4'-hydroxyacetophenone ≥98%
Bottle
Catalog # APOSBUP18685-1G
New Product
Supplier: Apollo Scientific
2-Bromo-4'-hydroxyacetophenone ≥98%
Catalog # APOSBUP18685-1G
Supplier: Apollo Scientific
New Product
CAS Number: 2491-38-5
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: 2-Bromo-1-(4-hydroxyphenyl)ethanone, 2-Bromo-4'-hydroxyacetophenone, 2-Bromo-1-(4-hydroxyphenyl)ethan-1-one, α-Bromo-4-hydroxyacetophenone, 4-Hydroxyphenacyl bromide
- CAS Number:2491-38-5
- Formula:C₈H₇BrO₂
- Molecular Weight:215,04 g/mol
Specifications
Specifications
- Pakning:Bottle
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



