Anisomycin ≥98%
Bottle
Katalog # APOSBUP17554-100MG
New Product
Leverandør: Apollo Scientific
Anisomycin ≥98%
Katalog # APOSBUP17554-100MG
Leverandør: Apollo Scientific
New Product
CAS nummer: 22862-76-6
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications
Synonymer: (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate
- CAS nummer:22862-76-6
- Formel:C₁₄H₁₉NO₄
- Molecular Weight:265,3 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle