Orders LinkContactUsLinkComponent
iFSP1 ≥98%
New Product
iFSP1 ≥98%
New Product
iFSP1 ≥98%
Usage: Biochemical/life science applications
  • Store at 2 to 8 °C
  • Non-hazardous
 :  1-amino-3-(4-methylphenyl)-pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile, 1-Amino-3-p-tolyl-benzo[4,5]imidazo[1,2-a]pyridine-2,4-dicarbonitrile
  •  :150651-39-1
  •  :C₂₀H₁₃N₅
  •  :MFCD01572665
  •  :323,35 g/mol
  •  :Køleskab
  •  :851-482-1
Product Family Options
  • Pakning
  • Bottle
    1 * 25 mg
    1 * 25 mg - 922,00 kr
    922,00 kr
    1 * 25 mg
       922,00 kr
    New Product
    New Product
  • Bottle
    1 * 50 mg
    1 * 50 mg - 1.690,00 kr
    1.690,00 kr
    1 * 50 mg
       1.690,00 kr
    New Product
    New Product
  • Bottle
    1 * 100 mg
    1 * 100 mg - 3.170,00 kr
    3.170,00 kr
    1 * 100 mg
       3.170,00 kr
    New Product
    New Product
  • Bottle
    1 * 500 mg
    1 * 500 mg - 7.040,00 kr
    7.040,00 kr
    1 * 500 mg
       7.040,00 kr