Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Ac-Met-Ome 99%

Supplier:   Apollo Scientific
Ac-Met-Ome 99%
Supplier:  Apollo Scientific
Certificates

4 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:35671-83-1
  • Formula:C₈H₁₅NO₃S
  • MDL Number:MFCD00038235
  • Molecular Weight:205,27 g/mol
Product Family Options

Product Information

  • Pakning
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 165,00 kr
    165,00 kr
    1 * 1 g
       165,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 7.920,00 kr
    7.920,00 kr
    1 * 100 g
       7.920,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 3.140,00 kr
    3.140,00 kr
    1 * 25 g
       3.140,00 kr

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 648,00 kr
    648,00 kr
    1 * 5 g
       648,00 kr