Orders LinkContactUsLinkComponent
Formamide ≥98%
Formamide ≥98%
Bottle
Katalog # APOSBUP19924-100G
New Product
Leverandør:   Apollo Scientific
Formamide ≥98%
Katalog # APOSBUP19924-100G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  75-12-7
Formamide ≥98%
Bottle
CAS nummer:  75-12-7
Price
396,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonymer:  Carbamaldehyde
  • CAS nummer:75-12-7
  • Formel:CH₃NO
  • Molecular Weight:45,04 g/mol
Specifikationer
Specifikationer
  • Pakning:
    Bottle