Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sodium iodide ≥98%

Sodium iodide ≥98%

Bottle
Katalog # APOSBUP15870-50MG
Irriterende
Miljø
Sundhed
New Product
Leverandør:   Apollo Scientific
Sodium iodide ≥98%
Katalog # APOSBUP15870-50MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  7681-82-5
Sodium iodide ≥98%

Bottle

CAS nummer:  7681-82-5
Price
286,00 kr
Log ind for tilgængelighed
1 * 50 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonymer:  Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
  • CAS nummer:7681-82-5
  • Formel:INa
  • MDL nummer:MFCD00003532
  • Molecular Weight:149,89 g/mol
  • EINECS:231-679-3
  • FN:3077
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.