3-AP ≥95%
New Product
Leverandør: Apollo Scientific
Gå til:
Om denne vare
Usage: Biochemical/ life science applications.
Synonymer: 3-AP, NSC 663249, 2-((3-Aminopyridin-2-yl)methylene)hydrazine-1-carbothioamide, 2-[(3-Aminopyridin-2-yl)methylene]hydrazinecarbothioamide, PAN-811, 3-Aminopyridine-2-Carboxyaldehyde Thiosemicarbazone
- CAS nummer:143621-35-6
- Formel:C₇H₉N₅S
- Molecular Weight:195,24 g/mol
4 tilgængelige muligheder nedenfor
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonymer: 3-AP, NSC 663249, 2-((3-Aminopyridin-2-yl)methylene)hydrazine-1-carbothioamide, 2-[(3-Aminopyridin-2-yl)methylene]hydrazinecarbothioamide, PAN-811, 3-Aminopyridine-2-Carboxyaldehyde Thiosemicarbazone
- CAS nummer:143621-35-6
- Formel:C₇H₉N₅S
- Molecular Weight:195,24 g/mol
Product Family Options
Produktinformation
- PakningTilgængelighedPrice
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



