Orders LinkContactUsLinkComponent
1-Tetradecanol
Bottle
  APOSBUP19399-1G
New Product
1-Tetradecanol
  APOSBUP19399-1G
New Product
 :  112-72-1
1-Tetradecanol
Bottle
 :  112-72-1
483,00 kr
1 * 1 g
Usage: Biochemical/ life science applications.
 :  1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol
  •  :112-72-1
  •  :C₁₄H₃₀O
  •  :214,39 g/mol
  • Pakning:
    Bottle