Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Water, purified

Water, purified

Bottle
Katalog # APOSBIP1406-10L
Leverandør:   Apollo Scientific
Water, purified
Katalog # APOSBIP1406-10L
Leverandør:  Apollo Scientific
CAS nummer:  7732-18-5
Water, purified

Bottle

CAS nummer:  7732-18-5
Price
209,00 kr
Log ind for tilgængelighed
1 * 10 L
Product Details & Documents

Usage: Biochemical/ life science applications.

  • Non-hazardous
Synonymer:  Hydrogen oxide
  • CAS nummer:7732-18-5
  • Formel:H₂O
  • MDL nummer:MFCD00011332
  • Molecular Weight:18,01528 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer