Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2-Chlorohexane 98%

New Product
2-Chlorohexane 98%
New Product
 
2-Chlorohexane 98%

 

 

Usage: Organic building block/reagent.

 :  2-Hexyl chloride
  •  :638-28-8
  •  :C₆H₁₃Cl
  •  :120,62 g/mol
 

 

Product Family Options

 

  • Pakning
     
     
  • Bottle
       
       
    1 * 1 g
    1 * 1 g - 275,00 kr
    275,00 kr
    1 * 1 g
       275,00 kr

     
    New Product
     
    Specifications
    Cat. No.
    APOSOR1023669-1G
    New Product
  • Bottle
       
       
    1 * 5 g
    1 * 5 g - 922,00 kr
    922,00 kr
    1 * 5 g
       922,00 kr

     
    New Product
     
    Specifications
    Cat. No.
    APOSOR1023669-5G
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.