Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phthalic acid ≥98%

Phthalic acid ≥98%

New Product
Leverandør:   Apollo Scientific
Phthalic acid ≥98%
Leverandør:  Apollo Scientific
New Product
Certifikater
Phthalic acid ≥98%

2 tilgængelige muligheder nedenfor

Product Details & Documents

Usage: Biochemical/life science applications.

Synonymer:  Pathalic acid
  • CAS nummer:88-99-3
  • Formel:C₈H₆O₄
  • Molecular Weight:166,13 g/mol
Product Family Options

Produktinformation

  • Pakning
    Tilgængelighed
    Price
  • Bottle
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 g
    1 * 5 g - 472,00 kr
    472,00 kr
    1 * 5 g
       472,00 kr

    Specifikationer:
    New Product
    New Product
  • Bottle
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 g
    1 * 10 g - 681,00 kr
    681,00 kr
    1 * 10 g
       681,00 kr

    Specifikationer:
    New Product
    New Product
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.