Biotin Hydrazide ≥95%
Bottle
Katalog # APOSBUP16575-100MG
New Product
Leverandør: Apollo Scientific
Biotin Hydrazide ≥95%
Katalog # APOSBUP16575-100MG
Leverandør: Apollo Scientific
New Product
CAS nummer: 66640-86-6
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonymer: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide
- CAS nummer:66640-86-6
- Formel:C₁₀H₁₈N₄O₂S
- Molecular Weight:258,34 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



